Product Details

SNP ID
rs6665599
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:247712281 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAGGCCACTGATGAGGGCTGTGG[C/G]CACTGCAATGGCCACGAAACCACAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR6F1 PubMed Links

Gene Details

Gene
OR6F1
Gene Name
olfactory receptor family 6 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005286.1 475 Missense Mutation CCC,GCC P159A NP_001005286.1

View Full Product Details