Product Details

SNP ID
rs112586220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27839049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACACCAGTTCCAAGTGAGCCCGC[C/T]CACCGCGGAACGTTCGGTCAGTCTC
Phenotype
MIM: 602788 MIM: 602801 MIM: 602825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AK PubMed Links

Gene Details

Gene
HIST1H2AK
Gene Name
histone cluster 1, H2ak
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003510.2 Intron NP_003501.1
Gene
HIST1H2BN
Gene Name
histone cluster 1, H2bn
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003520.3 Intron NP_003511.1
Gene
HIST1H4K
Gene Name
histone cluster 1, H4k
There are no transcripts associated with this gene.

View Full Product Details