Product Details

SNP ID
hCV25648792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24694045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGCGGGGCTCCGGGTTGGGACA[C/G]TGGCATGATTGCCGGTGGTGGGGGT
Phenotype
MIM: 312040
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POLA1 PubMed Links

Gene Details

Gene
POLA1
Gene Name
DNA polymerase alpha 1, catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016937.3 Intron NP_058633.2

View Full Product Details