Product Details

SNP ID
rs61730520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50333900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCGCGGTACTGCGCCTGCGCGG[C/T]CTCGCGCCTTCCCAGGTCCGCTCCG
Phenotype
MIM: 176264
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNC3 PubMed Links

Gene Details

Gene
KCNC3
Gene Name
potassium voltage-gated channel subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004977.2 Intron NP_004968.2
XM_006723203.2 Intron XP_006723266.1
XM_011526925.2 Intron XP_011525227.1
XM_011526926.1 Intron XP_011525228.1
XM_011526928.2 Intron XP_011525230.1
Gene
NAPSB
Gene Name
napsin B aspartic peptidase, pseudogene
There are no transcripts associated with this gene.

View Full Product Details