Product Details

SNP ID
rs6823412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139266831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTACTTCCGTGGTTTCCCTCGCC[A/G]CTGCATCTGTGACCTCTGGCCCGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MGARP PubMed Links

Gene Details

Gene
MGARP
Gene Name
mitochondria localized glutamic acid rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032623.3 671 Missense Mutation GCG,GTG A164V NP_116012.2

View Full Product Details