Product Details

SNP ID
rs61731352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143356904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCCAAGATAGGATCCTGTGGC[C/T]GGAGAGGCAATGAGGGACTGATCGC
Phenotype
MIM: 118425
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCN1 PubMed Links

Gene Details

Gene
CLCN1
Gene Name
chloride voltage-gated channel 1
There are no transcripts associated with this gene.

Gene
FAM131B
Gene Name
family with sequence similarity 131 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031690.2 865 Silent Mutation NP_001026860.2
NM_001278297.1 865 Silent Mutation NP_001265226.1
NM_014690.4 865 Silent Mutation NP_055505.3
XM_005250073.3 865 Silent Mutation XP_005250130.1
XM_006716186.2 865 Silent Mutation XP_006716249.1

View Full Product Details