Product Details

SNP ID
rs12307411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10825643 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGTCTCTCAATCCTGTCACATT[C/T]GATTGCATCTGCCTGTTGTGTCTCC
Phenotype
MIM: 604791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS2R10 PubMed Links

Gene Details

Gene
TAS2R10
Gene Name
taste 2 receptor member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023921.1 627 Silent Mutation TCA,TCG S209S NP_076410.1

View Full Product Details