Product Details

SNP ID
rs6778597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49676320 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCAGACCTGGGGAGGCACTAGCTT[C/T]TAGGAAGCAGAGCAGGTCCTATAGA
Phenotype
MIM: 102645 MIM: 604020 MIM: 142408
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APEH PubMed Links

Gene Details

Gene
APEH
Gene Name
acylaminoacyl-peptide hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001640.3 Intron NP_001631.3
XM_005265097.2 Intron XP_005265154.1
XM_005265098.4 Intron XP_005265155.1
XM_011533658.2 Intron XP_011531960.1
XM_011533659.1 Intron XP_011531961.1
XM_011533660.2 Intron XP_011531962.1
XM_011533661.2 Intron XP_011531963.1
XM_011533662.2 Intron XP_011531964.1
XM_011533663.2 Intron XP_011531965.1
XM_017006285.1 Intron XP_016861774.1
XM_017006286.1 Intron XP_016861775.1
Gene
BSN
Gene Name
bassoon presynaptic cytomatrix protein
There are no transcripts associated with this gene.

Gene
MST1
Gene Name
macrophage stimulating 1
There are no transcripts associated with this gene.

View Full Product Details