Product Details

SNP ID
rs61746548
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:3492708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCACTGACAGGGTCGCAGTCAG[C/T]CCCACCCCCGCAGTCACAGTGCAGG
Phenotype
MIM: 604266
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEGF6 PubMed Links

Gene Details

Gene
MEGF6
Gene Name
multiple EGF like domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001409.3 4309 Missense Mutation ACT,GCT T1483A NP_001400.3
XM_006710406.3 4309 Missense Mutation ACT,GCT T1378A XP_006710469.1
XM_011540885.1 4309 Missense Mutation ACT,GCT T1545A XP_011539187.1
XM_011540886.1 4309 Missense Mutation ACT,GCT T1502A XP_011539188.1
XM_011540887.2 4309 Missense Mutation ACT,GCT T1337A XP_011539189.1
XM_011540888.2 4309 Intron XP_011539190.1
XM_017000533.1 4309 Missense Mutation ACT,GCT T1501A XP_016856022.1

View Full Product Details