Product Details
- SNP ID
-
hCV25746580
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:152648641 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAAAAGACTATTTTATGATGTAT[A/G]TCAACAAAATTCAACCCCACTTTCT
- Phenotype
-
MIM: 600375
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
XRCC2
PubMed Links
Gene Details
- Gene
- XRCC2
- Gene Name
- X-ray repair cross complementing 2
There are no transcripts associated with this gene.
View Full Product Details