Product Details

SNP ID
rs3751334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95433756 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCCGCTGGCTGTGACGCTTGTCA[A/G]AGTGTTCTCTATCGGCTGCATGCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLDN10 PubMed Links

Gene Details

Gene
CLDN10
Gene Name
claudin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160100.1 158 UTR 5 NP_001153572.1
NM_006984.4 158 Intron NP_008915.1
NM_182848.3 158 UTR 5 NP_878268.1
XM_011521134.2 158 Intron XP_011519436.1
XM_017020843.1 158 UTR 5 XP_016876332.1
XM_017020844.1 158 UTR 5 XP_016876333.1

View Full Product Details