Product Details

SNP ID
rs61736194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95433769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACGCTTGTCAAAGTGTTCTCTAT[C/T]GGCTGCATGCCTAGACCACCAAAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN10 PubMed Links

Gene Details

Gene
CLDN10
Gene Name
claudin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160100.1 171 UTR 5 NP_001153572.1
NM_006984.4 171 Intron NP_008915.1
NM_182848.3 171 UTR 5 NP_878268.1
XM_011521134.2 171 Intron XP_011519436.1
XM_017020843.1 171 UTR 5 XP_016876332.1
XM_017020844.1 171 UTR 5 XP_016876333.1

View Full Product Details