Product Details

SNP ID
rs16861629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187121427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGACAAGAAAGAGGATCTTTAG[A/T]GCCACAGCGAGGGCTCATTGCCACT
Phenotype
MIM: 607547
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RPL39L PubMed Links

Gene Details

Gene
RPL39L
Gene Name
ribosomal protein L39 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052969.1 Intron NP_443201.1

View Full Product Details