Product Details

SNP ID
rs80020850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102082104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTTTAACATTACATAGTATAA[G/T]GCTTACCCAGACCAAGTTTGATCAT
Phenotype
MIM: 140571
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HSP90AA1 PubMed Links

Gene Details

Gene
HSP90AA1
Gene Name
heat shock protein 90 alpha family class A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017963.2 Intron NP_001017963.2
NM_005348.3 Intron NP_005339.3
XM_011536718.2 Intron XP_011535020.1

View Full Product Details