Product Details

SNP ID
rs377162265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:99222396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCCCTTCCATGATCCCCGCCT[C/T]TCCCAGAAGCCCTGACTCCTCCTGC
Phenotype
MIM: 607905 MIM: 609214
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALG2 PubMed Links

Gene Details

Gene
ALG2
Gene Name
ALG2, alpha-1,3/1,6-mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033087.3 Intron NP_149078.1
Gene
SEC61B
Gene Name
Sec61 translocon beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006808.2 Intron NP_006799.1

View Full Product Details