Product Details
- SNP ID
-
hCV25751758
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:74951562 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCTAGACTGAGCCCCTCGTCGG[C/G]TTCATCCTCACACCCGCTGTATCTC
- Phenotype
-
MIM: 605752
MIM: 607828
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HID1
PubMed Links
Gene Details
- Gene
- HID1
- Gene Name
- HID1 domain containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030630.2 |
2524 |
UTR 3 |
|
|
NP_085133.1 |
- Gene
- OTOP3
- Gene Name
- otopetrin 3
There are no transcripts associated with this gene.
View Full Product Details