Product Details

SNP ID
rs9900092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:51631572 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTTCACTGAGGTGGGATTCTTCT[G/T]GGCTTTGTTCCCTACTTGAGGAGCC
Phenotype
MIM: 604642
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CA10 PubMed Links

Gene Details

Gene
CA10
Gene Name
carbonic anhydrase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082533.1 1985 UTR 3 NP_001076002.1
NM_001082534.1 1985 UTR 3 NP_001076003.1
NM_020178.4 1985 UTR 3 NP_064563.1
XM_017024878.1 1985 UTR 3 XP_016880367.1

View Full Product Details