Product Details

SNP ID
rs9322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:17942399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCAATACAGCTCTGCAAAAGGGA[A/G]ACATTGTTCTGTGGGGAAAAAAGGC
Phenotype
MIM: 605937
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNX5 PubMed Links

Gene Details

Gene
SNX5
Gene Name
sorting nexin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282454.1 1497 Silent Mutation GTC,GTT V286V NP_001269383.1
NM_014426.3 1497 Silent Mutation GTC,GTT V391V NP_055241.1
NM_152227.2 1497 Silent Mutation GTC,GTT V391V NP_689413.1

View Full Product Details