Product Details

SNP ID
rs61740508
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:51290599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCGGCTGCTTCCCAATCACGGC[A/G]TAGCCGTTCCAGATGTACATCATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC39A PubMed Links

Gene Details

Gene
TTC39A
Gene Name
tetratricopeptide repeat domain 39A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080494.3 1500 Silent Mutation NP_001073963.1
NM_001144832.2 1500 Silent Mutation NP_001138304.1
NM_001297662.1 1500 Intron NP_001284591.1
NM_001297663.1 1500 Silent Mutation NP_001284592.1
NM_001297664.1 1500 Silent Mutation NP_001284593.1
NM_001297665.1 1500 Silent Mutation NP_001284594.1
NM_001297666.1 1500 Silent Mutation NP_001284595.1
NM_001297667.1 1500 Silent Mutation NP_001284596.1
XM_005270644.2 1500 Silent Mutation XP_005270701.1
XM_006710471.2 1500 Silent Mutation XP_006710534.1
XM_011541048.2 1500 Silent Mutation XP_011539350.1
XM_011541049.2 1500 Silent Mutation XP_011539351.1
XM_011541050.2 1500 Silent Mutation XP_011539352.1
XM_011541051.2 1500 Silent Mutation XP_011539353.1
XM_011541052.2 1500 Silent Mutation XP_011539354.1
XM_011541053.2 1500 Silent Mutation XP_011539355.1
XM_011541054.2 1500 Nonsense Mutation XP_011539356.1
XM_017000715.1 1500 Silent Mutation XP_016856204.1

View Full Product Details