Product Details

SNP ID
rs4800780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:21535935 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGGGAATTCTTGCATTAAAACTG[A/T]TCTTTACATGAAATGGAGACAAATT
Phenotype
MIM: 609674
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ESCO1 PubMed Links

Gene Details

Gene
ESCO1
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052911.2 Intron NP_443143.2
XM_011525798.1 Intron XP_011524100.1
XM_011525799.2 Intron XP_011524101.1
Gene
GREB1L
Gene Name
growth regulation by estrogen in breast cancer 1 like
There are no transcripts associated with this gene.

View Full Product Details