Product Details

SNP ID
rs61736667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:32630284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCATCATCTTCTCCTTCAGATA[C/T]AAGCAAATCCTCATACAGGACACTG
Phenotype
MIM: 607798
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF1L PubMed Links

Gene Details

Gene
TAF1L
Gene Name
TATA-box binding protein associated factor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153809.2 5386 Missense Mutation ATA,GTA I1766V NP_722516.1

View Full Product Details