Product Details

SNP ID
rs76752300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102846067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTGACAGCTGCTGAGACGTGTT[C/G]CAGCCAGAGTCTCTCCGCTTTAATG
Phenotype
MIM: 133530
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BIVM PubMed Links

Gene Details

Gene
BIVM
Gene Name
basic, immunoglobulin-like variable motif containing
There are no transcripts associated with this gene.

Gene
BIVM-ERCC5
Gene Name
BIVM-ERCC5 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204425.1 227 Intron NP_001191354.1
Gene
ERCC5
Gene Name
ERCC excision repair 5, endonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000123.3 227 UTR 5 NP_000114.2

View Full Product Details