Product Details

SNP ID
rs114861732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43955691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACGCTCTCGTGGCTGCGCCCAGG[G/T]TTCACCGGGAGAGAGGGTGCCAAGG
Phenotype
MIM: 609567 MIM: 612058
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PNPLA3 PubMed Links

Gene Details

Gene
PNPLA3
Gene Name
patatin like phospholipase domain containing 3
There are no transcripts associated with this gene.

Gene
SAMM50
Gene Name
SAMM50 sorting and assembly machinery component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015380.4 Intron NP_056195.3

View Full Product Details