Product Details

SNP ID
rs75972163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:141224347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTATTTTAAGCATGGGTAGGTGCT[C/T]GTATACATGGGTGGAGTTGGGGCTT
Phenotype
MIM: 609550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF330 PubMed Links

Gene Details

Gene
ZNF330
Gene Name
zinc finger protein 330
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292002.1 Intron NP_001278931.1
NM_014487.5 Intron NP_055302.1
XM_017008033.1 Intron XP_016863522.1

View Full Product Details