Product Details
- SNP ID
-
hCV25761992
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:30475462 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGGGGAAACTGAGGCTGGCCCCA[G/C]CCCACCTAGACTGCCTGAGCTCCTC
- Phenotype
-
MIM: 153370
- Polymorphism
- G/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ITGAL
PubMed Links
Gene Details
- Gene
- ITGAL
- Gene Name
- integrin subunit alpha L
View Full Product Details