Product Details

SNP ID
rs10110624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10725693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTCCTCTGCCACTCAAGGCACA[A/G]GAAGGAGAGGGCGCGAGGGCTGACC
Phenotype
MIM: 612202
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102723313 PubMed Links

Gene Details

Gene
LOC102723313
Gene Name
uncharacterized LOC102723313
There are no transcripts associated with this gene.

Gene
SOX7
Gene Name
SRY-box 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031439.3 1353 UTR 3 NP_113627.1

View Full Product Details