Product Details

SNP ID
rs76975306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113373937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCCCCAGGATGCCCTCTAGCC[A/G]TCGGTCAAAGTTGGAGATCACTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BSPRY PubMed Links

Gene Details

Gene
BSPRY
Gene Name
B-box and SPRY domain containing
There are no transcripts associated with this gene.

Gene
HDHD3
Gene Name
haloacid dehalogenase like hydrolase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304509.1 989 Missense Mutation CGG,TGG R140W NP_001291438.1
NM_001304510.1 989 Missense Mutation CGG,TGG R140W NP_001291439.1
NM_001304511.1 989 Missense Mutation CGG,TGG R140W NP_001291440.1
NM_031219.3 989 Missense Mutation CGG,TGG R140W NP_112496.1

View Full Product Details