Product Details
- SNP ID
-
rs3103220
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:42398092 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGGCGCTTTGCCTTCTCCTTGG[C/T]TGGCTGCGCTGGGGCCCGGCGGGCG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105374428
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11733156] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105374428
- Gene Name
- uncharacterized LOC105374428
There are no transcripts associated with this gene.
- Gene
- SHISA3
- Gene Name
- shisa family member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080505.1 |
254 |
Silent Mutation |
GGC,GGT |
G12G |
NP_001073974.1 |
View Full Product Details