Product Details

SNP ID
rs61733270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:26625398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCAGAGCCCCAAGGAGCAGAAC[A/G]GGGATTCCCTGTGCGCAGGCTCCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC3B PubMed Links

Gene Details

Gene
LRRC3B
Gene Name
leucine rich repeat containing 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317808.1 318 Intron NP_001304737.1
NM_001317809.1 318 Intron NP_001304738.1
NM_001317810.1 318 UTR 5 NP_001304739.1
NM_001317811.1 318 Intron NP_001304740.1
NM_052953.3 318 Intron NP_443185.1
XM_005264849.3 318 Intron XP_005264906.1
XM_005264850.4 318 Intron XP_005264907.1
XM_011533332.2 318 Intron XP_011531634.1
XM_017005675.1 318 UTR 5 XP_016861164.1

View Full Product Details