Product Details

SNP ID
rs7210156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38841208 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATCAGTTTTCCGCAAAAGGGGC[A/G]GAACCACGTGACTCTGGAAGTAGCT
Phenotype
MIM: 603662
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf98 PubMed Links

Gene Details

Gene
C17orf98
Gene Name
chromosome 17 open reading frame 98
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080465.2 182 Missense Mutation CCG,CTG P61L NP_001073934.1
Gene
RPL23
Gene Name
ribosomal protein L23
There are no transcripts associated with this gene.

View Full Product Details