Product Details

SNP ID
rs17523495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34840383 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTAGGTGACCATAGTTTTGATGA[C/T]CCAGGAACTGTAAATGTGCTTGCTT
Phenotype
MIM: 606586 MIM: 616344
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAI14 PubMed Links

Gene Details

Gene
RAI14
Gene Name
retinoic acid induced 14
There are no transcripts associated with this gene.

Gene
TTC23L
Gene Name
tetratricopeptide repeat domain 23 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317949.1 Intron NP_001304878.1
NM_144725.3 Intron NP_653326.3
XM_011513984.2 Intron XP_011512286.1
XM_011513985.2 Intron XP_011512287.1
XM_011513987.2 Intron XP_011512289.1
XM_017009119.1 Intron XP_016864608.1
XM_017009120.1 Intron XP_016864609.1
XM_017009121.1 Intron XP_016864610.1
XM_017009122.1 Intron XP_016864611.1
XM_017009123.1 Intron XP_016864612.1
XM_017009124.1 Intron XP_016864613.1
XM_017009125.1 Intron XP_016864614.1
XM_017009126.1 Intron XP_016864615.1

View Full Product Details