Product Details

SNP ID
hCV25772135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50559529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGGGTCTGGAGAACAGCTAGG[A/G]CCTGGCTCAGCCCTCTTGTGTCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf18 PubMed Links

Gene Details

Gene
C3orf18
Gene Name
chromosome 3 open reading frame 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171740.2 1198 UTR 3 NP_001165211.1
NM_001171741.2 1198 UTR 3 NP_001165212.1
NM_001171743.2 1198 UTR 3 NP_001165214.2
NM_016210.4 1198 UTR 3 NP_057294.2
Gene
HEMK1
Gene Name
HemK methyltransferase family member 1
There are no transcripts associated with this gene.

View Full Product Details