Product Details

SNP ID
rs3743623
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:83807955 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CGGGAGAAACTACTACTCCCATAAT[G/T]CCCCGCGGTCCCGCGAGCTGCCAGT
Phenotype
MIM: 604553
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HSBP1 PubMed Links

Gene Details

Gene
HSBP1
Gene Name
heat shock factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001537.3 53 UTR 5 NP_001528.1
Gene
LOC102724163
Gene Name
uncharacterized LOC102724163
There are no transcripts associated with this gene.

View Full Product Details