Product Details

SNP ID
rs3761090
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:35658548 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGTGAGGAAGATAAGAAGTCCATC[T/C]GTTCAGTTTCCCCACTGCGGAGGGA
Phenotype
MIM: 124089 MIM: 611557
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
COX6B1 PubMed Links

Gene Details

Gene
COX6B1
Gene Name
cytochrome c oxidase subunit 6B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001863.4 Intron NP_001854.1
Gene
UPK1A
Gene Name
uroplakin 1A
There are no transcripts associated with this gene.

Gene
UPK1A-AS1
Gene Name
UPK1A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details