Product Details

SNP ID
rs2237903
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:2946671 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGGAAATCACATTTAAGGGATGGC[C/T]GAAAAAATATGTGTAAAATATGCAA
Phenotype
MIM: 601651
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAP1L4 PubMed Links

Gene Details

Gene
NAP1L4
Gene Name
nucleosome assembly protein 1 like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005969.3 Intron NP_005960.1

View Full Product Details