Product Details

SNP ID
rs3769506
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:79136237 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTTCCTCCACATTTTCTCTAAGCT[T/G]CTTCTTTCCTGGAATGTTCACCTAC
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
REG1CP PubMed Links

Gene Details

Gene
REG1CP
Gene Name
regenerating family member 1 gamma, pseudogene
There are no transcripts associated with this gene.

View Full Product Details