Product Details

SNP ID
rs3775317
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:165333022 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGCTATTGCTCATTTAAATTAGAA[C/G]CTTTTCCATGAGAACCACACTGACG
Phenotype
MIM: 605774 MIM: 607545
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLHL2 PubMed Links

Gene Details

Gene
KLHL2
Gene Name
kelch like family member 2
There are no transcripts associated with this gene.

Gene
MSMO1
Gene Name
methylsterol monooxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017369.2 Intron NP_001017369.1
NM_006745.4 Intron NP_006736.1
XM_005263176.2 Intron XP_005263233.1

View Full Product Details