Product Details

SNP ID
rs114648302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18210820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTTGATTGTCTCCCTAAATGG[A/G]TGGGAAAGTGAAGCAGGAGCCACAT
Phenotype
MIM: 600128 MIM: 179490
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102725254 PubMed Links

Gene Details

Gene
LOC102725254
Gene Name
uncharacterized LOC102725254
There are no transcripts associated with this gene.

Gene
PDE4C
Gene Name
phosphodiesterase 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011528056.2 2218 UTR 3 XP_011526358.1
XM_011528058.2 2218 UTR 3 XP_011526360.1
XM_017026866.1 2218 UTR 3 XP_016882355.1
Gene
RAB3A
Gene Name
RAB3A, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details