Product Details

SNP ID
rs5973792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30218796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCCCAACCACTGCCGCTGCTGC[A/G]GCTGCGGGTGTTTCATCCACAAAAT
Phenotype
MIM: 300098
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEB2 PubMed Links

Gene Details

Gene
MAGEB2
Gene Name
MAGE family member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002364.4 359 Silent Mutation GCA,GCG A72A NP_002355.2
XM_011545512.1 359 Silent Mutation GCA,GCG A72A XP_011543814.1

View Full Product Details