Product Details

SNP ID
rs3093683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28339821 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATTCAGTCACTTTTCCTCTTCAC[C/T]TTATGTAGGGTGTCTTCACCTGGAG
Phenotype
MIM: 614394 MIM: 611519 MIM: 191161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT20 PubMed Links

Gene Details

Gene
IFT20
Gene Name
intraflagellar transport 20
There are no transcripts associated with this gene.

Gene
POLDIP2
Gene Name
DNA polymerase delta interacting protein 2
There are no transcripts associated with this gene.

Gene
TNFAIP1
Gene Name
TNF alpha induced protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021137.4 Intron NP_066960.1
XM_017024993.1 Intron XP_016880482.1

View Full Product Details