Product Details

SNP ID
rs61732615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51322101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCCTGGAGTTCAACTCTCCTGC[C/T]GACAACTACACAGTGTGTGAAGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGLON5 PubMed Links

Gene Details

Gene
IGLON5
Gene Name
IgLON family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101372.1 117 Silent Mutation GCC,GCT A39A NP_001094842.1
Gene
VSIG10L
Gene Name
V-set and immunoglobulin domain containing 10 like
There are no transcripts associated with this gene.

View Full Product Details