Product Details
- SNP ID
-
rs6569834
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:132726885 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAATTGAAACTCATCCACCACCACC[A/G]CAGCTGGGTCATGAGAAAAAATGTC
- Phenotype
-
MIM: 606592
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VNN3
PubMed Links
Gene Details
- Gene
- VNN3
- Gene Name
- vanin 3
View Full Product Details