Product Details

SNP ID
rs16859261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112123584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGATCGGGCATGCCTGGGGTC[C/T]GTAGAAGGCATATGTAGAAGTGAAT
Phenotype
MIM: 611956 MIM: 607792
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf52 PubMed Links

Gene Details

Gene
C3orf52
Gene Name
chromosome 3 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171747.1 528 Intron NP_001165218.1
NM_024616.2 528 Intron NP_078892.2
XM_011513133.2 528 Intron XP_011511435.1
XM_017007181.1 528 Intron XP_016862670.1
Gene
GCSAM
Gene Name
germinal center associated signaling and motility
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190259.1 528 Silent Mutation ACA,ACG T138T NP_001177188.1
NM_001190260.1 528 Silent Mutation ACA,ACG T121T NP_001177189.1
NM_152785.4 528 Silent Mutation ACA,ACG T136T NP_689998.1
XM_005247272.2 528 Silent Mutation ACA,ACG T119T XP_005247329.1
XM_011512627.2 528 Silent Mutation ACA,ACG T139T XP_011510929.1
XM_011512628.2 528 Silent Mutation ACA,ACG T137T XP_011510930.1
XM_011512629.1 528 Silent Mutation ACA,ACG T120T XP_011510931.1
XM_011512630.1 528 Silent Mutation ACA,ACG T120T XP_011510932.1

View Full Product Details