Product Details

SNP ID
rs75163084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169795468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGAAAAAGCCTTCAGTGAAGGA[A/G]AAAACTTAAAACTTACTATACACGT
Phenotype
MIM: 606042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC34 PubMed Links

Gene Details

Gene
LRRC34
Gene Name
leucine rich repeat containing 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172779.1 Intron NP_001166250.1
NM_001172780.1 Intron NP_001166251.1
NM_153353.4 Intron NP_699184.2
XM_006713508.3 Intron XP_006713571.1
XM_011512442.2 Intron XP_011510744.1
XM_017005746.1 Intron XP_016861235.1
Gene
MYNN
Gene Name
myoneurin
There are no transcripts associated with this gene.

View Full Product Details