Product Details

SNP ID
rs61738673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41217702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGAGAACAGTGATGATGTTTTT[A/G]GAAGTGTTGTAGATTTCACACCAGT
Phenotype
MIM: 609716
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TREML3P PubMed Links

Gene Details

Gene
TREML3P
Gene Name
triggering receptor expressed on myeloid cells like 3, pseudogene
There are no transcripts associated with this gene.

View Full Product Details