Product Details
- SNP ID
-
rs61741375
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:168057221 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTGCCAGCCTGCAGTCCAGGGCG[C/T]GGTGGAAGGTATGGCTGAGTGAGGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FRMD1
PubMed Links
Gene Details
- Gene
- FRMD1
- Gene Name
- FERM domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001122841.2 |
1441 |
Missense Mutation |
CAC,CGC |
H441R |
NP_001116313.1 |
NM_024919.4 |
1441 |
Missense Mutation |
CAC,CGC |
H509R |
NP_079195.3 |
XM_011536137.1 |
1441 |
Missense Mutation |
CAC,CGC |
H574R |
XP_011534439.1 |
XM_011536138.1 |
1441 |
Missense Mutation |
CAC,CGC |
H512R |
XP_011534440.1 |
XM_011536140.1 |
1441 |
Missense Mutation |
CAC,CGC |
H485R |
XP_011534442.1 |
XM_011536141.1 |
1441 |
Missense Mutation |
CAC,CGC |
H466R |
XP_011534443.1 |
XM_011536142.2 |
1441 |
Missense Mutation |
CAC,CGC |
H466R |
XP_011534444.1 |
XM_011536143.1 |
1441 |
Missense Mutation |
CAC,CGC |
H466R |
XP_011534445.1 |
XM_011536145.1 |
1441 |
Missense Mutation |
CAC,CGC |
H464R |
XP_011534447.1 |
XM_011536146.2 |
1441 |
Missense Mutation |
CAC,CGC |
H303R |
XP_011534448.1 |
XM_017011317.1 |
1441 |
Missense Mutation |
CAC,CGC |
H551R |
XP_016866806.1 |
XM_017011318.1 |
1441 |
Missense Mutation |
CAC,CGC |
H303R |
XP_016866807.1 |
View Full Product Details