Product Details
- SNP ID
-
rs916428342
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:54283490 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAAACCTCACAGCGAATGCGGGG[C/T]GCCACAGAAGCGACTTGGGGAGGGC
- Phenotype
-
MIM: 612985
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IRX3
PubMed Links
Gene Details
- Gene
- IRX3
- Gene Name
- iroquois homeobox 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024336.2 |
2118 |
UTR 3 |
|
|
NP_077312.2 |
View Full Product Details