Product Details

SNP ID
rs61746971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14820043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCAGCTCTTGAATGAGATTCAC[A/G]TAGTCAGTTTTTTCTGCACAGGCCC
Phenotype
MIM: 611233
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDNF PubMed Links

Gene Details

Gene
CDNF
Gene Name
cerebral dopamine neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029954.2 539 Silent Mutation TAC,TAT Y167Y NP_001025125.2
XM_011519488.2 539 Silent Mutation TAC,TAT Y176Y XP_011517790.1

View Full Product Details