Product Details

SNP ID
rs1046514093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:217799928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGCATACACCAGGATTTATACA[C/T]GGTGGCAGCGGCTATAGGCACGATG
Phenotype
MIM: 600076
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNS1 PubMed Links

Gene Details

Gene
TNS1
Gene Name
tensin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308022.1 9886 UTR 3 NP_001294951.1
NM_001308023.1 9886 UTR 3 NP_001294952.1
NM_022648.5 9886 UTR 3 NP_072174.3

View Full Product Details