Product Details

SNP ID
rs58662373
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:202169392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGCCTCATGGGGTGATGCCCC[C/T]GGGAAGCAGGATGGCTACTGCCTTC
Phenotype
MIM: 176889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPN7 PubMed Links

Gene Details

Gene
PTPN7
Gene Name
protein tyrosine phosphatase, non-receptor type 7
There are no transcripts associated with this gene.

Gene
PTPRVP
Gene Name
protein tyrosine phosphatase, receptor type V, pseudogene
There are no transcripts associated with this gene.

View Full Product Details